2-Bromo-5-ethylthiophene ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

A551621-5G A551621-250MG A551621-25G
77613-736EA 45.32 USD
77613-736 77684-395 77891-109
2-Bromo-5-ethylthiophene ≥96%, Ambeed.Inc
2-Bromo-5-ethylthiophene

Precaución: For research use only.

Formula: C₆H₇BrS
MDL Number: MFCD12033305
CAS Number: 62323-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR