Methyl 1-hydroxycyclohexanecarboxylate ≥95%

Supplier: Ambeed
Warning

A551192-1G A551192-5G
77686-415EA 75 USD
77686-415 77686-416
Methyl 1-hydroxycyclohexanecarboxylate ≥95%
Methyl 1-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD00182643
CAS Number: 6149-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR