1,4-Bis(4-bromophenyl)-1,3-butadiyne 95+%

Supplier: Ambeed
Warning

A543965-250MG A543965-1G A543965-5G
77686-401EA 44 USD
77686-401 77901-869 77900-937
1,4-Bis(4-bromophenyl)-1,3-butadiyne 95+%
1,4-Bis(4-bromophenyl)-1,3-butadiyne

Precaución: for Research Use Only

Formula: C₁₆H₈Br₂
MDL Number: MFCD00825678
CAS Number: 959-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR