8-Fluoroquinoline-7-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (8-Fluoroquinolin-7-yl)boronic acid

A543780-1G A543780-250MG A543780-5G
77882-317EA 354 USD
77882-317 77882-318 77882-319
8-Fluoroquinoline-7-boronic acid ≥98%
8-Fluoroquinoline-7-boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BFNO₂
MW: 190.970 g/mol
MDL Number: MFCD12025987
CAS Number: 1217500-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR