2-Amino-6-(2-(cyclopropylmethoxy)-6-hydroxyphenyl)-4-(piperidin-4-yl)nicotinonitrile hydrochloride ≥99%
Supplier: Ambeed
|
Warning
|
Synonyms:
ACHP HCl
A543661-1MG
A543661-10MG
A543661-5MG
77613-708EA
77.25
USD
77613-708
77684-385
77684-386
2-Amino-6-(2-(cyclopropylmethoxy)-6-hydroxyphenyl)-4-(piperidin-4-yl)nicotinonitrile hydrochloride ≥99%
2-Amino-6-(2-(cyclopropylmethoxy)-6-hydroxyphenyl)-4-(piperidin-4-yl)nicotinonitrile hydrochloride
Precaución: For research use only.
Formula:
C₂₁H₂₅ClN₄O₂ MW: 400.9 g/mol |
MDL Number:
MFCD22124458 CAS Number: 406209-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...