Silver(II) fluoride ≥99%

Supplier: Ambeed
Danger

A543632-5G A543632-1G A543632-250MG
77572-726EA 66 USD
77572-726 77669-402 77669-404
Silver(II) fluoride ≥99%
Silver(II) fluoride

Precaución: for Research Use Only

Formula: AgF₂
MDL Number: MFCD00003411
CAS Number: 7783-95-1
UN: 3084
ADR: 8,5.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR