3,5-Dichloropicolinaldehyde ≥96%

Supplier: Ambeed
Warning

A543515-100MG A543515-1G A543515-250MG
77872-907EA 14 USD
77872-907 77872-908 77872-909
3,5-Dichloropicolinaldehyde ≥96%
3,5-Dichloropicolinaldehyde

Precaución: for Research Use Only

Formula: C6H3Cl2NO
MDL Number: MFCD09839149
CAS Number: 471909-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR