2-Bromo-5-fluoro-4-methoxypyridine ≥98%

Supplier: Ambeed
Danger

A543375-1G A543375-100MG A543375-250MG
77613-702EA 116.39 USD
77613-702 77884-346 77884-347
2-Bromo-5-fluoro-4-methoxypyridine ≥98%
2-Bromo-5-fluoro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD18257621
CAS Number: 1256816-36-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR