(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3R,4R)-3-Fluoro-4-hydroxypyrrolidine-1-carboxylate

A541746-1G A541746-5G
77686-390EA 227 USD
77686-390 77686-391
(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate ≥98%
(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₆FNO₃
MW: 205.23 g/mol
MDL Number: MFCD19443950
CAS Number: 1174020-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR