(2-Methoxy-3-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-3-methylphenylboronic acid

A541572-1G A541572-100MG A541572-250MG A541572-5G
77673-448EA 108 USD
77673-448 77884-343 77884-344 77884-345
(2-Methoxy-3-methylphenyl)boronic acid ≥98%
(2-Methoxy-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MDL Number: MFCD06801731
CAS Number: 909187-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR