2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: [(tert-butoxycarbonyl)amino](3-chlorophenyl)acetic acid

A542666-1G A542666-250MG A542666-5G
77613-698EA 13.39 USD
77613-698 77686-396 77686-397
2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid ≥98%
2-((tert-Butoxycarbonyl)amino)-2-(3-chlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO₄
MDL Number: MFCD03426365
CAS Number: 669713-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR