Emedastine difumarate ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Ethoxyethyl)-2-(4-methyl-1,4-diazepan-1-yl)-1H-benzo[d]imidazole difumarate, 1-(2-Ethoxyethyl)-2-(4-methyl-1-homopiperazinyl)benzimidazole Difumarate

A541362-1G A541362-250MG
77602-302EA 171.18 USD
77602-302 77627-114
Emedastine difumarate ≥98%
Emedastine difumarate

Precaución: for Research Use Only

Formula: C₁₇H₂₆N₄O
MW: 534.57 g/mol
MDL Number: MFCD00923844
CAS Number: 87233-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR