1,1'-(2,2-Dimethylpropane-1,3-diyl)bis(3,4-dichloro-1H-pyrrole-2,5-dione) ≥97%

Supplier: Ambeed
Warning

Synonyms: NSC 617145

A533499-10MG A533499-1MG A533499-5MG
77617-270EA 141.11 USD
77617-270 77617-272 77617-274
1,1'-(2,2-Dimethylpropane-1,3-diyl)bis(3,4-dichloro-1H-pyrrole-2,5-dione) ≥97%
1,1'-(2,2-Dimethylpropane-1,3-diyl)bis(3,4-dichloro-1H-pyrrole-2,5-dione)

Precaución: for Research Use Only

Formula: C₁₃H₁₀Cl₄N₂O₄
MW: 400.04 g/mol
MDL Number: MFCD28100812
CAS Number: 203115-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR