Methyl 3-amino-2-methylpropanoate hydrochloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A533302-250MG A533302-100MG
77684-376EA 149 USD
77684-376 77886-334
Methyl 3-amino-2-methylpropanoate hydrochloride ≥95%, Ambeed.Inc
Methyl 3-amino-2-methylpropanoate hydrochloride

Precaución: For research use only.

Formula: C₅H₁₂ClNO₂
MDL Number: MFCD16547718
CAS Number: 88512-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR