2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide sulfate ≥98%

Supplier: Ambeed
Warning

Synonyms: Minoxidil sulphate

A540552-100G A540552-25G A540552-5G
77627-102EA 113.3 USD
77627-102 77627-104 77627-106
2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide sulfate ≥98%
2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide sulfate

Precaución: for Research Use Only

Formula: C₉H₁₅N₅O₄S
MW: 289.31 g/mol
MDL Number: MFCD06248907
CAS Number: 210357-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR