Ethyl isocyanatoacetate ≥97%

Supplier: Ambeed
Danger

A532982-25G A532982-5G
77572-690EA 139 USD
77572-690 77686-376
Ethyl isocyanatoacetate ≥97%
Ethyl isocyanatoacetate

Precaución: for Research Use Only

Formula: C₅H₇NO₃
MDL Number: MFCD00002040
CAS Number: 2949-22-6
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR