4-(tert-Butoxycarbonylamino)benzylamine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminobenzylamine, 4-BOC protected, 4-(Boc-amino)benzylamine, tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
A539817-1G
A539817-5G
A539817-250MG
A539817-10G
A539817-100MG
A539817-25G
77572-718EA
29
USD
77572-718
77572-720
77602-298
77669-390
77880-169
77880-170
4-(tert-Butoxycarbonylamino)benzylamine ≥98%
4-(tert-Butoxycarbonylamino)benzylamine
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂O₂ MW: 222.29 g/mol |
MDL Number:
MFCD02183573 CAS Number: 220298-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...