(R)-Pyrrolidine-3-carboxylic acid hydrochloride ≥97%

Supplier: Ambeed
Warning

A532542-1G A532542-250MG A532542-5G
77613-670EA 40.17 USD
77613-670 77613-672 77613-674
(R)-Pyrrolidine-3-carboxylic acid hydrochloride ≥97%
(R)-Pyrrolidine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClNO₂
MW: 151.59 g/mol
MDL Number: MFCD09954283
CAS Number: 1202245-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR