4-Bromo-2,5-dimethylbenzaldehyde ≥95%

Supplier: Ambeed
Warning

A539561-1G A539561-5G
77594-660EA 54.33 USD
77594-660 77594-662
4-Bromo-2,5-dimethylbenzaldehyde ≥95%
4-Bromo-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.071 g/mol
MDL Number: MFCD16295236
CAS Number: 88111-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR