Fmoc-Glu(OcHx)-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-glutamic acid 5-cyclohexyl ester, Fmoc-Glu(OcHex)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(cyclohexyloxy)-5-oxopentanoic acid

A539290-25G A539290-1G
77613-696EA 203.19 USD
77613-696 77901-596
Fmoc-Glu(OcHx)-OH ≥98%
Fmoc-Glu(OcHx)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₉NO₆
MDL Number: MFCD00270538
CAS Number: 150047-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR