S-Adenosyl-L-methionine tosylate ≥98%

Supplier: Ambeed
Warning

A531731-250MG A531731-100MG A531731-1G A531731-5G
77613-666EA 42.23 USD
77613-666 77673-422 77673-424 77673-426
S-Adenosyl-L-methionine tosylate ≥98%
S-Adenosyl-L-methionine tosylate

Precaución: for Research Use Only

Formula: C₂₂H₃₀N₆O₈S₂
MDL Number: MFCD00063498
CAS Number: 52248-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR