Chroman-6-ylboronic acid 98+%

Supplier: Ambeed
Warning

A531692-1G A531692-250MG
77884-319EA 445 USD
77884-319 77884-320
Chroman-6-ylboronic acid 98+%
Chroman-6-ylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MW: 177.993 g/mol
MDL Number: MFCD12779349
CAS Number: 279261-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR