Guanosine 5'-(trihydrogen diphosphate) P'-D-mannopyranosyl ester disodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: GDP-D-mannose disodium

A531449-10MG A531449-50MG
77602-272EA 97.38 USD
77602-272 77669-362
Guanosine 5'-(trihydrogen diphosphate) P'-D-mannopyranosyl ester disodium salt ≥98%
Guanosine 5'-(trihydrogen diphosphate) P'-D-mannopyranosyl ester disodium salt

Precaución: for Research Use Only

Formula: C₁₆H₂₃N₅Na₂O₁₆P₂
MW: 649.30 g/mol
MDL Number: MFCD00799981
CAS Number: 103301-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR