3-Methylenepiperidin-2-one ≥95%

Supplier: Ambeed
Warning

A530887-250MG A530887-1G
77749-108EA 244 USD
77749-108 77749-107
3-Methylenepiperidin-2-one ≥95%
3-Methylenepiperidin-2-one

Precaución: for Research Use Only

Formula: C₆H₉NO
MW: 111.14 g/mol
MDL Number: MFCD13176083
CAS Number: 68074-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR