(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-1-yl)propanoic acid ≥95%

Supplier: Ambeed
Warning

A530752-250MG A530752-100MG
77669-356EA 57 USD
77669-356 77880-151
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-1-yl)propanoic acid ≥95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₃NO₄
MDL Number: MFCD03428028
CAS Number: 511272-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR