(S)-2,4-Diaminobutanoic acid hydrobromide ≥95%

Supplier: Ambeed
Warning

A537942-1G A537942-250MG A537942-25G A537942-5G
77882-304EA 65 USD
77882-304 77882-305 77882-306 77882-307
(S)-2,4-Diaminobutanoic acid hydrobromide ≥95%
(S)-2,4-Diaminobutanoic acid hydrobromide

Precaución: for Research Use Only

Formula: C₄H₁₁BrN₂O₂
MDL Number: MFCD07368664
CAS Number: 73143-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR