4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: Trimethyl((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethynyl)silane, 4,4,5,5-Tetramethyl-2-(4-trimethylsilanylethynyl-phenyl)-[1,3,2]dioxaborolane

A537802-100MG A537802-1G A537802-250MG A537802-5G
77900-594EA 26 USD
77900-594 77900-926 77901-790 77900-853
4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester ≥98%
4-[(Trimethylsilyl)ethynyl]benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₅BO₂Si
MDL Number: MFCD08706286
CAS Number: 870238-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR