Benzylhydrazine ≥97%

Supplier: Ambeed
Danger

A530306-1G A530306-5G
77686-368EA 50 USD
77686-368 77686-369
Benzylhydrazine ≥97%
Benzylhydrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00869518
CAS Number: 555-96-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR