Methyl-1-methylpyrrolidine-3-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methylpyrrolidine-3-carboxylate

A537500-1G A537500-100MG
77747-096EA 72 USD
77747-096 77747-095
Methyl-1-methylpyrrolidine-3-carboxylate ≥97%
Methyl-1-methylpyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
MDL Number: MFCD12028048
CAS Number: 34616-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR