2-(3-((R)-3-(3,4-Dimethoxyphenyl)-1-(((S)-1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carbonyl)oxy)propyl)phenoxy)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: AP1867

A530140-250MG A530140-10MG A530140-50MG
77572-688EA 1420.37 USD
77572-688 77686-366 77686-367
2-(3-((R)-3-(3,4-Dimethoxyphenyl)-1-(((S)-1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carbonyl)oxy)propyl)phenoxy)acetic acid ≥98%
2-(3-((R)-3-(3,4-Dimethoxyphenyl)-1-(((S)-1-((S)-2-(3,4,5-trimethoxyphenyl)butanoyl)piperidine-2-carbonyl)oxy)propyl)phenoxy)acetic acid

Precaución: for Research Use Only

Formula: C₃₈H₄₇NO₁₁
MW: 693.78 g/mol
MDL Number: MFCD31813776
CAS Number: 195514-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR