(1S,9S)-1-Amino-9-ethyl-5-fluoro-9-hydroxy-4-methyl-2,3,12,15-tetrahydrobenzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinoline-10,13(1H,9H)-dione methanesulfonate ≥98%

Supplier: Ambeed
Danger

Synonyms: Exatecan mesylate

A537376-100MG A537376-1G A537376-50MG A537376-5G A537376-250MG
77572-702EA 82.4 USD
77572-702 77572-704 77617-294 77617-296 77747-094
(1S,9S)-1-Amino-9-ethyl-5-fluoro-9-hydroxy-4-methyl-2,3,12,15-tetrahydrobenzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinoline-10,13(1H,9H)-dione methanesulfonate ≥98%
(1S,9S)-1-Amino-9-ethyl-5-fluoro-9-hydroxy-4-methyl-2,3,12,15-tetrahydrobenzo[de]pyrano[3',4':6,7]indolizino[1,2-b]quinoline-10,13(1H,9H)-dione methanesulfonate

Precaución: for Research Use Only

Formula: C₂₅H₂₆FN₃O₇S
MW: 531.55 g/mol
MDL Number: MFCD04113012
CAS Number: 169869-90-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR