3-(1-(tert-Butoxycarbonyl)azetidin-3-yl)propanoic acid ≥98%

Supplier: Ambeed
Warning

A530070-100MG A530070-1G A530070-250MG
77598-544EA 74.83 USD
77598-544 77598-546 77598-548
3-(1-(tert-Butoxycarbonyl)azetidin-3-yl)propanoic acid ≥98%
3-(1-(tert-Butoxycarbonyl)azetidin-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD28388690
CAS Number: 1521114-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR