[1,2,4]Triazolo[4,3-a]pyridin-7-ylboronic acid ≥95%

Supplier: Ambeed
Warning

A529790-100MG A529790-250MG A529790-5G A529790-1G
77749-102EA 159 USD
77749-102 77749-104 77749-105 77749-103
[1,2,4]Triazolo[4,3-a]pyridin-7-ylboronic acid ≥95%
[1,2,4]Triazolo[4,3-a]pyridin-7-ylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BN₃O₂
MDL Number: MFCD27921702
CAS Number: 2304748-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR