3-Methoxy-9H-carbazole ≥95%

Supplier: Ambeed
Warning

A529788-250MG A529788-5G A529788-1G
77602-256EA 58.43 USD
77602-256 77673-408 77673-406
3-Methoxy-9H-carbazole ≥95%
3-Methoxy-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO
MDL Number: MFCD00626045
CAS Number: 18992-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR