6,6'-Thiobis(2-(tert-butyl)-4-methylphenol) ≥97%

Supplier: Ambeed
Warning

A529580-5G A529580-1G
77572-686EA 82.4 USD
77572-686 77594-636
6,6'-Thiobis(2-(tert-butyl)-4-methylphenol) ≥97%
6,6'-Thiobis(2-(tert-butyl)-4-methylphenol)

Precaución: for Research Use Only

Formula: C₂₂H₃₀O₂S
MW: 358.53 g/mol
MDL Number: MFCD00026291
CAS Number: 90-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR