Fenobam ≥98%

Supplier: Ambeed
Danger

Synonyms: 1-(3-Chlorophenyl)-3-(1-methyl-4-oxo-4,5-dihydro-1H-imidazol-2-yl)urea

A529486-1MG A529486-5MG
77602-254EA 20.5 USD
77602-254 77669-344
Fenobam ≥98%
Fenobam

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₄O₂
MW: 266.7 g/mol
MDL Number: MFCD00868019
CAS Number: 57653-26-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR