(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl (Z)-2-methylbut-2-enoate ≥98%

Supplier: Ambeed
Warning

Synonyms: Decursinol angelate

A529270-100MG A529270-50MG
77747-084EA 419 USD
77747-084 77747-085
(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl (Z)-2-methylbut-2-enoate ≥98%
(S)-8,8-Dimethyl-2-oxo-7,8-dihydro-2H,6H-pyrano[3,2-g]chromen-7-yl (Z)-2-methylbut-2-enoate

Precaución: for Research Use Only

Formula: C₁₉H₂₀O₅
MW: 328.35 g/mol
MDL Number: MFCD06796687
CAS Number: 130848-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR