2-((3-(4-Hydroxy-3,5-dimethoxyphenyl)acryloyl)oxy)-N,N,N-trimethylethan-1-aminium ≥98%

Supplier: Ambeed
Danger

Synonyms: Sinapine

A536549-10MG A536549-1MG A536549-5MG
77617-284EA 49.44 USD
77617-284 77617-286 77617-288
2-((3-(4-Hydroxy-3,5-dimethoxyphenyl)acryloyl)oxy)-N,N,N-trimethylethan-1-aminium ≥98%
2-((3-(4-Hydroxy-3,5-dimethoxyphenyl)acryloyl)oxy)-N,N,N-trimethylethan-1-aminium

Precaución: for Research Use Only

Formula: C₁₆H₂₄NO₅
CAS Number: 18696-26-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR