3-Bromo-1H-pyrrole-2-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A528878-1G A528878-250MG
77594-634EA 177.33 USD
77594-634 77622-356
3-Bromo-1H-pyrrole-2-carboxylic acid ≥97%
3-Bromo-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂
MW: 189.9948 g/mol
MDL Number: MFCD28128036
CAS Number: 145821-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR