(E)-4-Ethoxy-1,1,1-trifluorobut-3-en-2-one ≥98%

Supplier: Ambeed
Danger

A528810-25G A528810-5G
77884-313EA 75 USD
77884-313 77884-314
(E)-4-Ethoxy-1,1,1-trifluorobut-3-en-2-one ≥98%
(E)-4-Ethoxy-1,1,1-trifluorobut-3-en-2-one

Precaución: for Research Use Only

Formula: C₆H₇F₃O₂
MDL Number: MFCD00192131
CAS Number: 59938-06-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR