Ethyl 6-bromoimidazo[1,2-a]pyrazine-3-carboxylate ≥97%

Supplier: Ambeed
Warning

A536365-1G A536365-250MG
77617-278EA 183.34 USD
77617-278 77747-093
Ethyl 6-bromoimidazo[1,2-a]pyrazine-3-carboxylate ≥97%
Ethyl 6-bromoimidazo[1,2-a]pyrazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrN₃O₂
MW: 270.08 g/mol
MDL Number: MFCD28347683
CAS Number: 1289193-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR