S-(((S)-2-Acetamido-3-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-amino-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropan-2-yl)amino)-3-oxopropyl)thio)-L-cysteine hydrochloride (1:X) ≥98%

Supplier: Ambeed
Warning

A536362-1MG A536362-5MG
77617-276EA 30.9 USD
77617-276 77749-119
S-(((S)-2-Acetamido-3-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-amino-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropan-2-yl)amino)-3-oxopropyl)thio)-L-cysteine hydrochloride (1:X) ≥98%
S-(((S)-2-Acetamido-3-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-(((R)-1-amino-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-5-guanidino-1-oxopentan-2-yl)amino)-1-oxopropa

Precaución: for Research Use Only

Formula: C₃₈H₇₄ClN₂₁O₁₀S₂
MDL Number: MFCD32004547
CAS Number: 1334237-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR