1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: Ambeed
Danger

A536202-5G A536202-1G
77572-698EA 47.38 USD
77572-698 77684-378
1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥97%
1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: For research use only.

Formula: C₁₀H₃F₉O
MW: 310.11 g/mol
MDL Number: MFCD22688445
CAS Number: 130336-17-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR