3H-Imidazo[4,5-b]pyridine-2-carboxylic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A535883-1G A535883-100MG
77684-377EA 540 USD
77684-377 77886-341
3H-Imidazo[4,5-b]pyridine-2-carboxylic acid ≥95%, Ambeed.Inc
3H-Imidazo[4,5-b]pyridine-2-carboxylic acid

Precaución: For research use only.

Formula: C₇H₅N₃O₂
MDL Number: MFCD05721719
CAS Number: 97640-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR