(R)-1-(5-Hydroxyhexyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione ≥97%
Supplier: Ambeed
|
Warning
|
A528014-5MG
A528014-10MG
A528014-1MG
77747-081EA
188
USD
77747-081
77747-079
77747-080
(R)-1-(5-Hydroxyhexyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione ≥97%
(R)-1-(5-Hydroxyhexyl)-3,7-dimethyl-1H-purine-2,6(3H,7H)-dione
Precaución: for Research Use Only
Formula:
C₁₃H₂₀N₄O₃ |
MDL Number:
MFCD00867573 CAS Number: 100324-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...