N-Benzylquininium chloride ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Benzylquinine chloride, QUIBEC, (1S,2S,4S,5R)-1-Benzyl-2-((R)-hydroxy(6-methoxyquinolin-4-yl)methyl)-5-vinylquinuclidin-1-ium chloride

A535867-1G A535867-5G A535867-25G
77602-282EA 13.33 USD
77602-282 77602-284 77669-374
N-Benzylquininium chloride ≥98%
N-Benzylquininium chloride

Precaución: for Research Use Only

Formula: C₂₇H₃₁ClN₂O₂
MW: 451.01 g/mol
MDL Number: MFCD00198105
CAS Number: 67174-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR