2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)methyl]benzeneboronic acid, pinacol ester, 2-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione
A535687-100MG
A535687-1G
A535687-250MG
A535687-5G
77880-159EA
11
USD
77880-159
77880-160
77880-161
77880-162
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester ≥98%
2-[(Phthalimid-1-yl)methyl]benzeneboronic acid, pinacol ester
Precaución: for Research Use Only
Formula:
C₂₁H₂₂BNO₄ |
MDL Number:
MFCD02179492 CAS Number: 380430-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...