Potassium trioxalatoferrate(III) trihydrate ≥97%

Supplier: Ambeed
Warning

Synonyms: Ferrate(3-), tris[ethanedioato(2-)-κO1,κO2]-, potassium, hydrate (1:3:3), (OC-6-11)-, Potassium iron(III) oxalate

A527735-100G A527735-1000G A527735-500G
77572-682EA 11 USD
77572-682 77669-334 77669-336
Potassium trioxalatoferrate(III) trihydrate ≥97%
Potassium trioxalatoferrate(III) trihydrate

Precaución: for Research Use Only

Formula: K₃Fe(C₂O₄)₃·3H₂O
MDL Number: MFCD00150450
CAS Number: 5936-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR