Quinoline-6-sulfonyl chloride ≥98%

Supplier: Ambeed
Danger

A535206-100MG A535206-1G
77872-895EA 142 USD
77872-895 77872-896
Quinoline-6-sulfonyl chloride ≥98%
Quinoline-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂S
MDL Number: MFCD03086095
CAS Number: 65433-99-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR