POPSO sesquisodium salt (β,β'-Dihydroxypiperazine-1,4-dipropanesulfonic acid sesquisodium salt) ≥98%

Supplier: Ambeed
Warning

Synonyms: POPSO sesquisodium, 3,3'-(Piperazine-1,4-diyl)bis(2-hydroxypropane-1-sulphonic acid), sodium salt(2:3)

A535192-25G A535192-5G A535192-100G
77602-278EA 39.98 USD
77602-278 77602-280 77669-372
POPSO sesquisodium salt (β,β'-Dihydroxypiperazine-1,4-dipropanesulfonic acid sesquisodium salt) ≥98%
POPSO sesquisodium salt (β,β'-Dihydroxypiperazine-1,4-dipropanesulphonic acid sesquisodium salt)

Precaución: for Research Use Only

Formula: C₂₀H₄₁N₄Na₃O₁₆S₄
MW: 793.8099 g/mol
MDL Number: MFCD00070004
CAS Number: 108321-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR